Methyl-4-(cyanomethyl)benzoate ≥95.0%

Supplier: TCI America

Synonyms: Methyl 4-(cyanomethyl)benzoate, 4-(Cyanomethyl)benzoic Acid Methyl Ester

M2341-25G M2341-5G
TCM2341-25GEA 407.64 CAD
TCM2341-25G TCM2341-5G
Methyl-4-(cyanomethyl)benzoate ≥95.0%
Methyl-4-(cyanomethyl)benzoate

CAS Number: 76469-88-0
MDL Number: MFCD00060695
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 62


Other Product Names / Synonyms: 4-(Cyanomethyl)benzoic Acid Methyl Ester

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 112 °C
Melting Pt: 62 °C
MDL Number: MFCD00060695
CAS Number: 76469-88-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49044d16e2b07d6ae352efb50466d9a85670e4a6&vid=49044d16e2b07d6ae352efb50466d9a85670e4a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR