1-Methyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0%

Supplier: TCI America

Synonyms: 1-Methylpyrazole-4-boronic acid pinacol ester, 1-Methyl-4-pyrazole boronic acid pinacol ester, 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

M2339-1G M2339-5G
TCM2339-1GEA 83.74 CAD
TCM2339-1G TCM2339-5G
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester ≥98.0%
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester

CAS Number: 761446-44-0
MDL Number: MFCD03789259
Molecular Formula: C10H17BN2O2
Molecular Weight: 208.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 1-Methylpyrazole-4-boronic Acid Pinacol Ester, 2-(1-Methyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Formula: C₁₀H₁₇BN₂O₂
MW: 208.07 g/mol
Melting Pt: 62 °C
MDL Number: MFCD03789259
CAS Number: 761446-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f4f480b78608e6d456f6182111aadeecbaf7bed&vid=4f4f480b78608e6d456f6182111aadeecbaf7bed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR