4-Maleimidobutyric acid ≥98.0%
Supplier: TCI America
Synonyms:
4-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)butanoic acid
M2337-1G
M2337-5G
TCM2337-1GEA
115.34
CAD
TCM2337-1G
TCM2337-5G
4-Maleimidobutyric acid ≥98.0%
4-Maleimidobutyric acid
CAS Number: 57078-98-5
MDL Number: MFCD00043139
Molecular Formula: C8H9NO4
Molecular Weight: 183.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 96
Storage Temperature: 0-10°C
Formula:
C₈H₉NO₄ MW: 183.16 g/mol Melting Pt: 96 °C |
MDL Number:
MFCD00043139 CAS Number: 57078-98-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...