2-Methoxy-5-pyrimidineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (2-Methoxypyrimidin-5-yl)boronic acid

M2335-1G M2335-5G
TCM2335-1GEA 67.94 CAD
TCM2335-1G TCM2335-5G
2-Methoxy-5-pyrimidineboronic acid (contains varying amounts of Anhydride)
2-Methoxy-5-pyrimidineboronic acid

CAS Number: 628692-15-9
MDL Number: MFCD03094664
Molecular Formula: C5H7BN2O3
Molecular Weight: 153.93
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 161


Other Product Names / Synonyms: 2-Methoxypyrimidine-5-boronic Acid, 2-Methoxy-5-pyrimidinylboronic Acid

Formula: C₅H₇BN₂O₃
MW: 153.9317 g/mol
Melting Pt: 161 °C
MDL Number: MFCD03094664
CAS Number: 628692-15-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89b830e2dc882b27717e4984a4d224d63ff22aaa&vid=89b830e2dc882b27717e4984a4d224d63ff22aaa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR