Methyl hydrogen isophthalate ≥98.0% (by GC)

Supplier: TCI America

M2327-1G M2327-5G
TCM2327-1GEA 36.34 CAD
TCM2327-1G TCM2327-5G
Methyl hydrogen isophthalate ≥98.0% (by GC)
Methyl hydrogen isophthalate

CAS Number: 1877-71-0
MDL Number: MFCD00029972
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 194


Other Product Names / Synonyms: Methyl Hydrogen Isophthalate, Isophthalic Acid Monomethyl Ester

Formula: C₉H₈O₄
Melting Pt: 194 °C
MDL Number: MFCD00029972
CAS Number: 1877-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91fc74587d788ec7552dfac3038809a0f9748e33&vid=91fc74587d788ec7552dfac3038809a0f9748e33&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR