4-(2-Methyl-4-thiazolyl)phenol ≥98.0% (by HPLC, total nitrogen) for biochemical research

Supplier: TCI America

M2325-100MG M2325-1G
TCM2325-100MGEA 90.06 CAD
TCM2325-100MG TCM2325-1G
4-(2-Methyl-4-thiazolyl)phenol ≥98.0% (by HPLC, total nitrogen) for biochemical research
4-(2-Methyl-4-thiazolyl)phenol

[for Biochemical Research]
CAS Number: 30686-73-8
MDL Number: MFCD02642878
Molecular Formula: C10H9NOS
Molecular Weight: 191.25
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 213

Formula: C₁₀H₉NOS
Melting Pt: 213 °C
MDL Number: MFCD02642878
CAS Number: 30686-73-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a06575890278672de463e455e623cb6cbfda3800&vid=faf6fbcdf020824c9e2c276768d56d90e5f1978e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR