Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 6-chloro-3,5-diaminopyrazine-2-carboxylate, Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate

M2322-25G M2322-5G
TCM2322-25GEA 370.19 CAD
TCM2322-25G TCM2322-5G
Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate ≥98.0%
Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate

CAS Number: 1458-01-1
MDL Number: MFCD01928388
Molecular Formula: C6H7ClN4O2
Molecular Weight: 202.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 213


Other Product Names / Synonyms: 3,5-Diamino-6-chloropyrazine-2-carboxylic Acid Methyl Ester

Formula: C₆H₇ClN₄O₂
Melting Pt: 213 °C
MDL Number: MFCD01928388
CAS Number: 1458-01-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0d6da6503b02d09a89f89bb902023944de77a8d&vid=f0d6da6503b02d09a89f89bb902023944de77a8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR