1-Methyl-3-[6-(methylthio)hexyl]imidazolium p-toluenesulfonate ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America

M2321-1G M2321-5G
TCM2321-1GEA 330.54 CAD
TCM2321-1G TCM2321-5G
1-Methyl-3-[6-(methylthio)hexyl]imidazolium p-toluenesulfonate ≥95.0% (by HPLC, total nitrogen)
1-Methyl-3-[6-(methylthio)hexyl]imidazolium p-toluenesulfonate

CAS Number: 1352947-63-7
Molecular Formula: C18H28N2O3S2
Molecular Weight: 384.55
Purity/Analysis Method: >95.0% (HPLC,N)
Form: Crystal

Formula: C₁₈H₂₈N₂O₃S₂
CAS Number: 1352947-63-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e65c93de6013ce4fe7994162d08a9331349f7ab2&vid=e65c93de6013ce4fe7994162d08a9331349f7ab2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR