Methyl-5-acetamido-7,8,9-tri-O-acetyl-5-N-4-O-carbonyl-3,5-dideoxy-2-S-phenyl-2-thio-D-glycero-β-D-galacto-2-nonulopyranosylonate ≥96.0% (by HPLC)

Supplier: TCI America

M2319-1G M2319-200MG
TCM2319-1GEA 1824.9 CAD
TCM2319-1G TCM2319-200MG
Methyl-5-acetamido-7,8,9-tri-O-acetyl-5-N-4-O-carbonyl-3,5-dideoxy-2-S-phenyl-2-thio-D-glycero-β-D-galacto-2-nonulopyranosylonate ≥96.0% (by HPLC)
Methyl-5-acetamido-7,8,9-tri-O-acetyl-5-N-4-O-carbonyl-3,5-dideoxy-2-S-phenyl-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosylonate

CAS Number: 934591-76-1
Molecular Formula: C25H29NO12S
Molecular Weight: 567.56
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Melting point (°C): 152
Specific rotation [a]20/D: -123 deg (C=0.8, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Glycosylation, Bioscience, Thioglycosides, Sialic Acids
Other Product Names / Synonyms: Methyl (Phenyl 5-Acetamido-7,8,9-tri-O-acetyl-5-N,4-O-carbonyl-3,5-dideoxy-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosid)onate

Formula: C₂₅H₂₉NO₁₂S
Melting Pt: 152 °C
MDL Number: MFCD22581442
CAS Number: 934591-76-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0623f4ecc833b02a7487528537e5ce407a898a8c&vid=0623f4ecc833b02a7487528537e5ce407a898a8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR