2'-O-Methylguanosine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Amino-9-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-1H-purin-6(9H)-one

M2318-1G M2318-200MG
TCM2318-1GEA 416.49 CAD
TCM2318-1G TCM2318-200MG
2'-O-Methylguanosine ≥98.0% (by HPLC, titration analysis)
2'-O-Methylguanosine

CAS Number: 2140-71-8
MDL Number: MFCD00057053
Molecular Formula: C11H15N5O5
Molecular Weight: 297.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 236
Specific rotation [a]20/D: -55 deg (C=1, 1mol/L NaOH)
Storage Temperature: 0-10°C

Formula: C₁₁H₁₅N₅O₅
MW: 297.27 g/mol
Melting Pt: 236 °C
MDL Number: MFCD00057053
CAS Number: 2140-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef53f8f6256375e44e6a5fa8da5e16c6b169eb59&vid=ef53f8f6256375e44e6a5fa8da5e16c6b169eb59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR