2'-O-Methylcytidine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
M2317-1G
M2317-200MG
TCM2317-1GEA
293.88
CAD
TCM2317-1G
TCM2317-200MG
2'-O-Methylcytidine ≥98.0% (by HPLC, titration analysis)
2'-O-Methylcytidine
CAS Number: 2140-72-9
MDL Number: MFCD00056067
Molecular Formula: C10H15N3O5
Molecular Weight: 257.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 257
Specific rotation [a]20/D: 72 deg (C=1, H2O)
Storage Temperature: 0-10°C
Formula:
C₁₀H₁₅N₃O₅ Melting Pt: 257 °C |
MDL Number:
MFCD00056067 CAS Number: 2140-72-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...