4-(Methylthio)benzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: 4-(Methylthio)benzyl Alcohol

M2314-25G M2314-5G
TCM2314-25GEA 951.16 CAD
TCM2314-25G TCM2314-5G
4-(Methylthio)benzyl alcohol ≥98.0%
4-(Methylthio)benzyl alcohol

CAS Number: 3446-90-0
MDL Number: MFCD00009706
Molecular Formula: C8H10OS
Molecular Weight: 154.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 109
Melting point (°C): 41
Flash Point (°C): 113

Formula: C₈H₁₀OS
MW: 154.23 g/mol
Boiling Pt: 109 °C
Melting Pt: 41 °C
Flash Pt: 113 °C
MDL Number: MFCD00009706
CAS Number: 3446-90-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b58fb769585375b34fa905a246c305437aa166bc&vid=b58fb769585375b34fa905a246c305437aa166bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR