2-Methoxy-5-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 2-Methoxy-5-methylbenzeneboronic acid

M2311-25G M2311-5G
TCM2311-25GEA 1834.38 CAD
TCM2311-25G TCM2311-5G
2-Methoxy-5-methylphenylboronic acid (contains varying amounts of Anhydride)
2-Methoxy-5-methylphenylboronic acid

CAS Number: 127972-00-3
MDL Number: MFCD02179465
Molecular Formula: C8H11BO3
Molecular Weight: 165.98
Form: Crystal
Color: White
Melting point (°C): 97


Other Product Names / Synonyms: 2-Methoxy-5-methylbenzeneboronic Acid

Formula: C₈H₁₁BO₃
MW: 165.99 g/mol
Melting Pt: 97 °C
MDL Number: MFCD02179465
CAS Number: 127972-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c73eb46ab0f244978d3e57adc9e5e21d837bb74&vid=0c73eb46ab0f244978d3e57adc9e5e21d837bb74&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR