Methyl trans-6-octadecenoate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: trans-6-Octadecenoic Acid Methyl Ester, Methyl Petroselaidate, Petroselaidic Acid Methyl Ester, Methyl trans-6-Octadecenoate

M2310-100MG M2310-500MG
TCM2310-100MGEA 204.93 CAD
TCM2310-100MG TCM2310-500MG
Methyl trans-6-octadecenoate ≥97.0% (by GC)
Methyl trans-6-octadecenoate

CAS Number: 14620-36-1
MDL Number: MFCD00069607
Molecular Formula: C19H36O2
Molecular Weight: 296.50
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Melting point (°C): 19
Storage Temperature: <0°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: trans-6-Octadecenoic Acid Methyl Ester, Methyl Petroselaidate, Petroselaidic Acid Methyl Ester

Formula: C₁₉H₃₆O₂
MW: 296.50 g/mol
Boiling Pt: 165 °C
Melting Pt: 19 °C
MDL Number: MFCD00069607
CAS Number: 14620-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e0c8ca6b7ac3cb5b21552586368e12777c875cc&vid=8e0c8ca6b7ac3cb5b21552586368e12777c875cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR