Methyl cis-11-octadecenoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl cis-Vaccenate, cis-Vaccenic Acid Methyl Ester, cis-11-Octadecenoic Acid Methyl Ester

M2308-100MG M2308-500MG
TCM2308-100MGEA 143.78 CAD
TCM2308-100MG TCM2308-500MG
Methyl cis-11-octadecenoate ≥98.0% (by GC)
Methyl cis-11-octadecenoate

CAS Number: 1937-63-9
MDL Number: MFCD00010457
Molecular Formula: C19H36O2
Molecular Weight: 296.50
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 62
Specific Gravity (20/20): 0.88
Storage Temperature: <0°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: cis-11-Octadecenoic Acid Methyl Ester, Methyl cis-Vaccenate, cis-Vaccenic Acid Methyl Ester

Formula: C₁₉H₃₆O₂
MW: 296.50 g/mol
Boiling Pt: 136 °C
Density: 0.88 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00010457
CAS Number: 1937-63-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9c1d6ebd99d7d4a50b0d54c7908a2b7c19d279f&vid=f9c1d6ebd99d7d4a50b0d54c7908a2b7c19d279f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR