Methyl trans,trans-9,12-octadecadienoate ≥98.0% (by GC)
Supplier: TCI America
M2307-100MG
M2307-500MG
TCM2307-100MGEA
74.26
CAD
TCM2307-100MG
TCM2307-500MG
Methyl trans,trans-9,12-octadecadienoate ≥98.0% (by GC)
Methyl trans,trans-9,12-octadecadienoate
CAS Number: 2566-97-4
MDL Number: MFCD00069997
Molecular Formula: C19H34O2
Molecular Weight: 294.48
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 152
Specific Gravity (20/20): 0.88
Storage Temperature: <0°C
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: trans,trans-9,12-Octadecadienoic Acid Methyl Ester, Methyl Linolelaidate, Linolelaidic Acid Methyl Ester
Formula:
C₁₉H₃₄O₂ Boiling Pt: 152 °C Density: 0.88 (20 °C) |
MDL Number:
MFCD00069997 CAS Number: 2566-97-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...