3-Methoxypyridine ≥96.0% (by GC, titration analysis)

Supplier: TCI America

M2304-1G M2304-5G
TCM2304-1GEA 41.08 CAD
TCM2304-1G TCM2304-5G
3-Methoxypyridine ≥96.0% (by GC, titration analysis)
3-Methoxypyridine

CAS Number: 7295-76-3
MDL Number: MFCD00673022
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 179
Flash Point (°C): 71
Specific Gravity (20/20): 1.08

Formula: C₆H₇NO
Boiling Pt: 179 °C
Density: 1.08 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00673022
CAS Number: 7295-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0f8955d6b22d290690147bfa0375ae8ed7d4d1b&vid=b0f8955d6b22d290690147bfa0375ae8ed7d4d1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR