Methyl-4-aminonicotinate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2303-1G
M2303-5G
TCM2303-1GEA
238.58
CAD
TCM2303-1G
TCM2303-5G
Methyl-4-aminonicotinate ≥98.0% (by GC, titration analysis)
Methyl 4-aminonicotinate
CAS Number: 16135-36-7
MDL Number: MFCD00955996
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 174
Other Product Names / Synonyms: 4-Aminonicotinic Acid Methyl Ester, Methyl 4-Aminopyridine-3-carboxylate, 4-Aminopyridine-3-carboxylic Acid Methyl Ester
Formula:
C₇H₈N₂O₂ Melting Pt: 174 °C |
MDL Number:
MFCD00955996 CAS Number: 16135-36-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...