Miglitol ≥98.0%
Supplier: TCI America
Synonyms:
Diastabol, BAY1099, Seibule, BAY-m1099, Glyset, (2R,3R,4R,5S)-1-(2-Hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol
M2302-1G
M2302-5G
TCM2302-1GEA
274.45
CAD
TCM2302-1G
TCM2302-5G
Miglitol ≥98.0%
Miglitol
CAS Number: 72432-03-2
MDL Number: MFCD00867240
Molecular Formula: C8H17NO5
Molecular Weight: 207.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 146
Specific rotation [a]20/D: -8 deg (C=0.5, H2O)
Formula:
C₈H₁₇NO₅ Melting Pt: 146 °C |
MDL Number:
MFCD00867240 CAS Number: 72432-03-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...