2-Mercapto-5-(trifluoromethyl)pyridine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M2299-25G M2299-5G
TCM2299-25GEA 586.18 CAD
TCM2299-25G TCM2299-5G
2-Mercapto-5-(trifluoromethyl)pyridine ≥98.0% (by HPLC, titration analysis)
2-Mercapto-5-(trifluoromethyl)pyridine

CAS Number: 76041-72-0
MDL Number: MFCD00128893
Molecular Formula: C6H4F3NS
Molecular Weight: 179.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 150
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Thiols, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-(Trifluoromethyl)-2-pyridinethiol, 5-(Trifluoromethyl)-2(1H)-pyridinethione

Formula: C₆H₄F₃NS
Melting Pt: 150 °C
MDL Number: MFCD00128893
CAS Number: 76041-72-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fd335ccb954c7de97ba7c8ffff9b42d5d689f18&vid=7fd335ccb954c7de97ba7c8ffff9b42d5d689f18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR