2-(4-Methylphenyl)ethanol ≥98.0% (by GC)

Supplier: TCI America

M2298-25G M2298-5G
TCM2298-25GEA 357.08 CAD
TCM2298-25G TCM2298-5G
2-(4-Methylphenyl)ethanol ≥98.0% (by GC)
2-(4-Methylphenyl)ethanol

CAS Number: 699-02-5
MDL Number: MFCD00002904
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 107
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 4-Methylphenethyl Alcohol, 2-(4-Methylphenyl)ethyl Alcohol, 2-(4-Methylphenyl)ethanol

Formula: C₉H₁₂O
Boiling Pt: 115 °C
Density: 1.00 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00002904
CAS Number: 699-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a8ef9394e7a624b98daadc1fd128bb00416c935&vid=5a8ef9394e7a624b98daadc1fd128bb00416c935&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR