5-Methoxy-1-tetralone ≥98.0%
Supplier: TCI America
Synonyms:
5-Methoxy-3,4-dihydronaphthalenone
M2297-5G
M2297-25G
TCM2297-5GEA
169.06
CAD
TCM2297-5G
TCM2297-25G
5-Methoxy-1-tetralone ≥98.0%
5-Methoxy-1-tetralone
CAS Number: 33892-75-0
MDL Number: MFCD00001692
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 162
Melting point (°C): 90
Other Product Names / Synonyms: 3,4-Dihydro-5-methoxy-1(2H)-naphthalenone
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol Boiling Pt: 162 °C Melting Pt: 90 °C |
MDL Number:
MFCD00001692 CAS Number: 33892-75-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...