2'-O-Methyluridine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 2′-O-Methyluridine
M2290-1G
M2290-5G
TCM2290-1GEA
104.28
CAD
TCM2290-1G
TCM2290-5G
2'-O-Methyluridine ≥98.0% (by HPLC, titration analysis)
2'-O-Methyluridine
CAS Number: 2140-76-3
MDL Number: MFCD00056054
Molecular Formula: C10H14N2O6
Molecular Weight: 258.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 161
Specific rotation [a]20/D: 42 deg (C=2, H2O)
Lambda max.: 261 nm (MeOH)Storage Temperature: 0-10°C
Formula:
C₁₀H₁₄N₂O₆ MW: 258.23 g/mol Melting Pt: 161 °C |
MDL Number:
MFCD00056054 CAS Number: 2140-76-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...