Monosodium risedronate hemipentahydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M2289-100MG M2289-1G
TCM2289-100MGEA 210.57 CAD
TCM2289-100MG TCM2289-1G
Monosodium risedronate hemipentahydrate ≥98.0% (by HPLC, titration analysis)
Monosodium risedronate hemipentahydrate

CAS Number: 329003-65-8
MDL Number: MFCD01706268
Molecular Formula: C7H11NO7P2
Molecular Weight: 305.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Anti-osteoporosis Ingredients
Other Product Names / Synonyms: Monosodium 1-Hydroxy-2-(3-pyridyl)ethylidene-1,1-diphosphonate, Risedronic Acid Monosodium Salt

Formula: C₇H₁₁NO₇P₂
MDL Number: MFCD01706268
CAS Number: 329003-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97849560c19e7c50c2fd80df3e51969cd3024b84&vid=97849560c19e7c50c2fd80df3e51969cd3024b84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR