4-Chloro-2-picoline ≥98.0% (by GC)
Supplier: TCI America
M2284-1G
M2284-5G
TCM2284-1GEA
60.04
CAD
TCM2284-1G
TCM2284-5G
4-Chloro-2-picoline ≥98.0% (by GC)
4-Chloro-2-picoline
CAS Number: 3678-63-5
MDL Number: MFCD00038833
Molecular Formula: C6H6ClN
Molecular Weight: 127.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 56
Specific Gravity (20/20): 1.15
Other Product Names / Synonyms: 4-Chloro-2-picoline
Formula:
C₆H₆ClN Boiling Pt: 56 °C Density: 1.15 (20 °C) |
MDL Number:
MFCD00038833 CAS Number: 3678-63-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...