4-Methoxy-3-nitropyridine ≥98.0% (by GC)
Supplier: TCI America
M2283-1G
M2283-5G
TCM2283-1GEA
134.3
CAD
TCM2283-1G
TCM2283-5G
4-Methoxy-3-nitropyridine ≥98.0% (by GC)
4-Methoxy-3-nitropyridine
CAS Number: 31872-62-5
MDL Number: MFCD00209661
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 127
Melting point (°C): 76
Formula:
C₆H₆N₂O₃ Boiling Pt: 127 °C Melting Pt: 76 °C |
MDL Number:
MFCD00209661 CAS Number: 31872-62-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...