DL-4-Methoxymandelic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Hydroxy-2-(4-methoxyphenyl)acetic acid

M2278-25G M2278-5G
TCM2278-25GEA 597.24 CAD
TCM2278-25G TCM2278-5G
DL-4-Methoxymandelic acid ≥98.0%
DL-4-Methoxymandelic acid

CAS Number: 10502-44-0
MDL Number: MFCD00004233
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 107

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Melting Pt: 107 °C
MDL Number: MFCD00004233
CAS Number: 10502-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94f0cb9afa8232153b2023a6b92b4e5457dfa94d&vid=94f0cb9afa8232153b2023a6b92b4e5457dfa94d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR