Mebendazole ≥98.0%

Supplier: TCI America

Synonyms: Methyl [5-(Benzoyl)benzimidazol-2-yl]carbamate, Methyl (5-benzoyl-1H-benzo[d]imidazol-2-yl)carbamate

M2273-25G M2273-5G
TCM2273-25GEA 107.44 CAD
TCM2273-25G TCM2273-5G
Mebendazole ≥98.0%
Mebendazole

CAS Number: 31431-39-7
MDL Number: MFCD00057872
Molecular Formula: C16H13N3O3
Molecular Weight: 295.30
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 287


Product Categories: Bioscience, Anthelmintic Ingredients
Other Product Names / Synonyms: Methyl [5-(Benzoyl)benzimidazol-2-yl]carbamate

Formula: C₁₆H₁₃N₃O₃
MW: 295.30 g/mol
Melting Pt: 287 °C
MDL Number: MFCD00057872
CAS Number: 31431-39-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c33fae3a9e728fae7ed4ab8265a48fc27a9c7693&vid=c33fae3a9e728fae7ed4ab8265a48fc27a9c7693&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR