1-Methylimidazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2272-1G M2272-5G
TCM2272-1GEA 127.98 CAD
TCM2272-1G TCM2272-5G
1-Methylimidazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)
1-Methylimidazole-4-carboxylic acid

CAS Number: 41716-18-1
MDL Number: MFCD02179560
Molecular Formula: C5H6N2O2
Molecular Weight: 126.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 247

Formula: C₅H₆N₂O₂
Melting Pt: 247 °C
MDL Number: MFCD02179560
CAS Number: 41716-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08d86b917bf01b40a40059362739b3736ba7abf2&vid=08d86b917bf01b40a40059362739b3736ba7abf2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR