3-Methylsalicylaldehyde ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2270-25G
M2270-5G
TCM2270-25GEA
509.08
CAD
TCM2270-25G
TCM2270-5G
3-Methylsalicylaldehyde ≥98.0% (by GC, titration analysis)
3-Methylsalicylaldehyde
CAS Number: 824-42-0
MDL Number: MFCD00040798
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 209
Melting point (°C): 17
Flash Point (°C): 87
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: 2-Hydroxy-3-methylbenzaldehyde
Formula:
C₈H₈O₂ Boiling Pt: 209 °C Melting Pt: 17 °C Density: 1.13 (20 °C) Flash Pt: 87 °C |
MDL Number:
MFCD00040798 CAS Number: 824-42-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...