4-(4-Methylpiperazin-1-yl)benzoic acid ≥98.0%
Supplier: TCI America
Synonyms:
1-(4-Carboxyphenyl)-4-methylpiperazine, 4-(4-Methylpiperazinyl)benzoic acid
M2267-1G
M2267-5G
TCM2267-1GEA
107.44
CAD
TCM2267-1G
TCM2267-5G
4-(4-Methylpiperazin-1-yl)benzoic acid ≥98.0%
4-(4-Methylpiperazin-1-yl)benzoic acid
CAS Number: 86620-62-4
MDL Number: MFCD02682063
Molecular Formula: C12H16N2O2
Molecular Weight: 220.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 230
Other Product Names / Synonyms: 1-(4-Carboxyphenyl)-4-methylpiperazine
Formula:
C₁₂H₁₆N₂O₂ MW: 220.2700 g/mol Melting Pt: 230 °C |
MDL Number:
MFCD02682063 CAS Number: 86620-62-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...