6-Mercapto-1-hexanol ≥98.0%

Supplier: TCI America

M2266-25G M2266-5G
TCM2266-25GEA 610.75 CAD
TCM2266-25G TCM2266-5G
6-Mercapto-1-hexanol ≥98.0%
6-Mercapto-1-hexanol

CAS Number: 1633-78-9
MDL Number: MFCD00068552
Molecular Formula: C6H14OS
Molecular Weight: 134.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 110
Specific Gravity (20/20): 0.99

Formula: C₆H₁₄OS
Boiling Pt: 105 °C
Density: 0.99 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00068552
CAS Number: 1633-78-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e25449096c01152c9ba1e52a3ff4f762fd1ac0d&vid=4e25449096c01152c9ba1e52a3ff4f762fd1ac0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR