Methyl-5-bromovalerate ≥97.0%

Supplier: TCI America

Synonyms: Methyl 5-Bromovalerate, 5-Bromovaleric acid methyl ester

M2265-25G M2265-5G
TCM2265-25GEA 249.64 CAD
TCM2265-25G TCM2265-5G
Methyl-5-bromovalerate ≥97.0%
Methyl 5-bromovalerate

CAS Number: 5454-83-1
MDL Number: MFCD00000265
Molecular Formula: C6H11BrO2
Molecular Weight: 195.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 89
Flash Point (°C): 99
Specific Gravity (20/20): 1.37


Other Product Names / Synonyms: 5-Bromovaleric Acid Methyl Ester

Formula: BrH₂C(CH₂)₃COOCH₃
MW: 195.06
Boiling Pt: 89 °C
Density: 1.37 (20 °C)
Flash Pt: 99 °C
MDL Number: MFCD00000265
CAS Number: 5454-83-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13a6e765b9b4864d5b23faded216516b6f31f6a0&vid=13a6e765b9b4864d5b23faded216516b6f31f6a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR