Methyl-9-formylnonanoate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 9-Formylnonanoic Acid Methyl Ester

M2263-1G M2263-5G
TCM2263-1GEA 409.22 CAD
TCM2263-1G TCM2263-5G
Methyl-9-formylnonanoate ≥95.0% (by GC)
Methyl-9-formylnonanoate

CAS Number: 14811-73-5
Molecular Formula: C11H20O3
Molecular Weight: 200.28
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Specific Gravity (20/20): 0.97
Storage Temperature: <0°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 9-Formylnonanoic Acid Methyl Ester

Formula: C₁₁H₂₀O₃
MW: 200.28 g/mol
Boiling Pt: 121 °C
Density: 0.97 (20 °C)
CAS Number: 14811-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=468e18f116df70b50272ccf662a17e469aec9092&vid=468e18f116df70b50272ccf662a17e469aec9092&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR