2-Methacryloyloxy-2-methyladamantane ≥97.0% stabilised

Supplier: TCI America

Synonyms: Methacrylic Acid 2-Methyl-2-adamantyl Ester, 2-Methyl-2-adamantyl Methacrylate, 2-Methyladamantan-2-yl methacrylate

M2260-5G M2260-1G
TCM2260-5GEA 342.86 CAD
TCM2260-5G TCM2260-1G
2-Methacryloyloxy-2-methyladamantane ≥97.0% stabilised
2-Methacryloyloxy-2-methyladamantane

(stabilized with MEHQ)
CAS Number: 177080-67-0
MDL Number: MFCD04114354
Molecular Formula: C15H22O2
Molecular Weight: 234.34
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 120
Specific Gravity (20/20): 1.06
Storage Temperature: <0°C


Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents
Other Product Names / Synonyms: 2-Methyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Methyl-2-adamantyl Ester

Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Density: 1.06 (20 °C)
Flash Pt: 120 °C
MDL Number: MFCD04114354
CAS Number: 177080-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab15e21b8289e0380d75ce355bc3e9fbb9ffe737&vid=ab15e21b8289e0380d75ce355bc3e9fbb9ffe737&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR