Methyl trans-p-coumarate ≥98.0% (by GC)
Supplier: TCI America
M2259-25G
M2259-5G
TCM2259-25GEA
360.24
CAD
TCM2259-25G
TCM2259-5G
Methyl trans-p-coumarate ≥98.0% (by GC)
Methyl trans-p-coumarate
CAS Number: 19367-38-5
MDL Number: MFCD00157167
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 139
Specific rotation [a]20/D: 12.1 deg (C=0.12, MeOH)
Lambda max.: 315 nm (MeOH)
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: trans-p-Coumaric Acid Methyl Ester, trans-4-Hydroxycinnamic Acid Methyl Ester, Methyl trans-4-Hydroxycinnamate
Formula:
C₁₀H₁₀O₃ Melting Pt: 139 °C |
MDL Number:
MFCD00157167 CAS Number: 19367-38-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...