3-(Methylaminomethyl)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-Methyl-1-(pyridin-3-yl)methanamine, N-Methyl-3-picolylamine, N-(3-Pyridylmethyl)methylamine

M2257-1G M2257-5G
TCM2257-1GEA 66.04 CAD
TCM2257-1G TCM2257-5G
3-(Methylaminomethyl)pyridine ≥98.0% (by GC, titration analysis)
3-(Methylaminomethyl)pyridine

CAS Number: 20173-04-0
MDL Number: MFCD00023610
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 93
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: N-Methyl-3-picolylamine, N-(3-Pyridylmethyl)methylamine

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 98 °C
Density: 1.01 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00023610
CAS Number: 20173-04-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e89da5213d47738fa5a263704e6e2c4c9cb9fbd&vid=9e89da5213d47738fa5a263704e6e2c4c9cb9fbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR