Methyl-3-hydroxy-4-nitrobenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3-hydroxy-4-nitrobenzoate

M2250-25G M2250-5G
TCM2250-25GEA 391.68 CAD
TCM2250-25G TCM2250-5G
Methyl-3-hydroxy-4-nitrobenzoate ≥98.0%
Methyl 3-hydroxy-4-nitrobenzoate

CAS Number: 713-52-0
MDL Number: MFCD00016997
Molecular Formula: C8H7NO5
Molecular Weight: 197.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 92


Other Product Names / Synonyms: 3-Hydroxy-4-nitrobenzoic Acid Methyl Ester

Formula: C₈H₇NO₅
Melting Pt: 92 °C
MDL Number: MFCD00016997
CAS Number: 713-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83797d53cbf755404cfc4b15c9676ccf0712a3c6&vid=83797d53cbf755404cfc4b15c9676ccf0712a3c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR