6-Methyl-4-chromanone ≥98.0% (by GC)

Supplier: TCI America

M2249-1G M2249-5G
TCM2249-1GEA 83.74 CAD
TCM2249-1G TCM2249-5G
6-Methyl-4-chromanone ≥98.0% (by GC)
6-Methyl-4-chromanone

CAS Number: 39513-75-2
MDL Number: MFCD00024071
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 162
Melting point (°C): 35

Formula: C₁₀H₁₀O₂
Boiling Pt: 162 °C
Melting Pt: 35 °C
MDL Number: MFCD00024071
CAS Number: 39513-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03bd8548a12845b8380500a7c56db67ddcdfec91&vid=03bd8548a12845b8380500a7c56db67ddcdfec91&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR