Methyl-4-bromo-3-hydroxybenzoate ≥98.0% (by GC)

Supplier: TCI America

M2246-1G M2246-5G
TCM2246-1GEA 97.96 CAD
TCM2246-1G TCM2246-5G
Methyl-4-bromo-3-hydroxybenzoate ≥98.0% (by GC)
Methyl 4-bromo-3-hydroxybenzoate

CAS Number: 106291-80-9
MDL Number: MFCD08056372
Molecular Formula: C8H7BrO3
Molecular Weight: 231.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 123


Other Product Names / Synonyms: 4-Bromo-3-hydroxybenzoic Acid Methyl Ester

Formula: C₈H₇BrO₃
Melting Pt: 123 °C
MDL Number: MFCD08056372
CAS Number: 106291-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=144f4e5246e9680a2094007b0d388c3a707af42c&vid=144f4e5246e9680a2094007b0d388c3a707af42c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR