6-Nitro-m-anisic acid ≥97.0%

Supplier: TCI America

Synonyms: 5-Methoxy-2-nitrobenzoic acid

M2245-25G M2245-5G
TCM2245-25GEA 213.3 CAD
TCM2245-25G TCM2245-5G
6-Nitro-m-anisic acid ≥97.0%
6-Nitro-m-anisic acid

CAS Number: 1882-69-5
MDL Number: MFCD00151836
Molecular Formula: C8H7NO5
Molecular Weight: 197.15
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 132


Other Product Names / Synonyms: 6-Nitro-m-anisic Acid

Formula: C₈H₇NO₅
Melting Pt: 132 °C
MDL Number: MFCD00151836
CAS Number: 1882-69-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaddc35c1faa53f1acfbd3840de80f60378f9b58&vid=eaddc35c1faa53f1acfbd3840de80f60378f9b58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR