4-Methoxy-2,6-dimethylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
M2244-1G
M2244-5G
TCM2244-1GEA
128.93
CAD
TCM2244-1G
TCM2244-5G
4-Methoxy-2,6-dimethylphenylboronic acid (contains varying amounts of Anhydride)
4-Methoxy-2,6-dimethylphenylboronic acid
CAS Number: 361543-99-9
MDL Number: MFCD03452755
Molecular Formula: C9H13BO3
Molecular Weight: 180.01
Form: Crystal
Color: White
Melting point (°C): 175
Other Product Names / Synonyms: 4-Methoxy-2,6-dimethylbenzeneboronic Acid
Formula:
C₉H₁₃BO₃ Melting Pt: 175 °C |
MDL Number:
MFCD03452755 CAS Number: 361543-99-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...