1-Methyl-4-piperidinemethanol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

M2242-25G M2242-5G
TCM2242-25GEA 407.64 CAD
TCM2242-25G TCM2242-5G
1-Methyl-4-piperidinemethanol ≥97.0% (by GC, titration analysis)
1-Methyl-4-piperidinemethanol

CAS Number: 20691-89-8
MDL Number: MFCD03411903
Molecular Formula: C7H15NO
Molecular Weight: 129.20
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 110
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 4-Hydroxymethyl-1-methylpiperidine

Formula: C₇H₁₅NO
Boiling Pt: 108 °C
Density: 0.97 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD03411903
CAS Number: 20691-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e36683170f35865373a13ac9bb5b21f4941cee1&vid=1e36683170f35865373a13ac9bb5b21f4941cee1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR