Methyl-2-chloro-6-methylisonicotinate ≥97.0% (by GC)

Supplier: TCI America

M2239-1G M2239-5G
TCM2239-1GEA 52.14 CAD
TCM2239-1G TCM2239-5G
Methyl-2-chloro-6-methylisonicotinate ≥97.0% (by GC)
Methyl 2-chloro-6-methylisonicotinate

CAS Number: 3998-90-1
MDL Number: MFCD04038391
Molecular Formula: C8H8ClNO2
Molecular Weight: 185.61
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 60


Other Product Names / Synonyms: 2-Chloro-6-methylisonicotinic Acid Methyl Ester, 2-Chloro-6-methylpyridine-4-carboxylic Acid Methyl Ester, Methyl 2-Chloro-6-methylpyridine-4-carboxylate

Formula: C₈H₈ClNO₂
Boiling Pt: 125 °C
Melting Pt: 60 °C
MDL Number: MFCD04038391
CAS Number: 3998-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74dfc1e05883eb3c5e81e40b920747577d8feff3&vid=74dfc1e05883eb3c5e81e40b920747577d8feff3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR