7-Methoxycoumarin-3-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 7-Methoxy-2-oxo-2H-chromene-3-carboxylic acid

M2233-100MG M2233-1G
TCM2233-100MGEA 45.82 CAD
TCM2233-100MG TCM2233-1G
7-Methoxycoumarin-3-carboxylic acid ≥98.0% (by GC, titration analysis)
7-Methoxycoumarin-3-carboxylic acid

CAS Number: 20300-59-8
MDL Number: MFCD00452770
Molecular Formula: C11H8O5
Molecular Weight: 220.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 196
Storage Temperature: <0°C

Formula: C₁₁H₈O₅
MW: 220.18 g/mol
Melting Pt: 196 °C
MDL Number: MFCD00452770
CAS Number: 20300-59-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac08078d4e66b93da43157f51425b27166c02377&vid=ac08078d4e66b93da43157f51425b27166c02377&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR