Methyl-1H-imidazole-5-carboxylate ≥97.0% (by GC, titration analysis)
Supplier: TCI America
M2228-1G
M2228-5G
TCM2228-1GEA
42.66
CAD
TCM2228-1G
TCM2228-5G
Methyl-1H-imidazole-5-carboxylate ≥97.0% (by GC, titration analysis)
Methyl-1H-imidazole-5-carboxylate
CAS Number: 17325-26-7
MDL Number: MFCD00216584
Molecular Formula: C5H6N2O2
Molecular Weight: 126.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 153
Other Product Names / Synonyms: 1H-Imidazole-5-carboxylic Acid Methyl Ester
Formula:
C₅H₆N₂O₂ Melting Pt: 153 °C |
MDL Number:
MFCD00216584 CAS Number: 17325-26-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...