1-Boc-isonipecotic acid methyl ester ≥98.0%

Supplier: TCI America

Synonyms: Methyl piperidine-4-carboxylate, N-BOC protected, 1-tert-Butyl 4-methyl piperidine-1,4-dicarboxylate

M2227-1G M2227-5G
TCM2227-1GEA 31.6 CAD
TCM2227-1G TCM2227-5G
1-Boc-isonipecotic acid methyl ester ≥98.0%
1-Boc-isonipecotic acid methyl ester

CAS Number: 124443-68-1
MDL Number: MFCD02183584
Molecular Formula: C12H21NO4
Molecular Weight: 243.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 35


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)

Formula: C₁₂H₂₁NO₄
MW: 243.2994 g/mol
Melting Pt: 35 °C
MDL Number: MFCD02183584
CAS Number: 124443-68-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19fcda41c1f1ea6c5565b57ac39a15d9037c32a3&vid=19fcda41c1f1ea6c5565b57ac39a15d9037c32a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR