1-Methyl-1H-pyrazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2224-1G M2224-5G
TCM2224-1GEA 161.95 CAD
TCM2224-1G TCM2224-5G
1-Methyl-1H-pyrazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)
1-Methyl-1H-pyrazole-4-carboxylic acid

CAS Number: 5952-92-1
MDL Number: MFCD00159641
Molecular Formula: C5H6N2O2
Molecular Weight: 126.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 207

Formula: C₅H₆N₂O₂
MW: 126.1100 g/mol
Melting Pt: 207 °C
MDL Number: MFCD00159641
CAS Number: 5952-92-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58331c438386982c4f4a5aa486b755149ff0d4b2&vid=58331c438386982c4f4a5aa486b755149ff0d4b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR