Methyl-2-methyl-5-nitrobenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-methyl-5-nitrobenzoate

M2220-25G M2220-5G
TCM2220-25GEA 171.9 CAD
TCM2220-25G TCM2220-5G
Methyl-2-methyl-5-nitrobenzoate ≥98.0%
Methyl 2-methyl-5-nitrobenzoate

CAS Number: 77324-87-9
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 69


Other Product Names / Synonyms: 2-Methyl-5-nitrobenzoic Acid Methyl Ester, Methyl 5-Nitro-o-toluate, 5-Nitro-o-toluic Acid Methyl Ester

Formula: C₉H₉NO₄
Melting Pt: 69 °C
MDL Number: MFCD00230865
CAS Number: 77324-87-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=516303c06ea0db21483f7676c3d9227e900ad8c8&vid=516303c06ea0db21483f7676c3d9227e900ad8c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR