2-Methyl-m-anisic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Methoxy-2-methylbenzoic acid

M2217-25G M2217-5G
TCM2217-25GEA 105.86 CAD
TCM2217-25G TCM2217-5G
2-Methyl-m-anisic acid ≥98.0% (by HPLC, titration analysis)
2-Methyl-m-anisic acid

CAS Number: 55289-06-0
MDL Number: MFCD02094039
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 149


Other Product Names / Synonyms: 3-Methoxy-o-toluic Acid, 2-Methyl-m-anisic Acid

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 149 °C
MDL Number: MFCD02094039
CAS Number: 55289-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c896ae6565a786a0e6677f9b435891cc1866f89d&vid=c896ae6565a786a0e6677f9b435891cc1866f89d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR